EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Crystal structure of glycoside hydrolase family 127 β-l-arabinofuranosidase from Bifidobacterium longum.
PMID 24680821
Abstract
Enzymes acting on β-linked arabinofuranosides have been unknown until recently, in spite of wide distribution of β-l-arabinofuranosyl oligosaccharides in plant cells. Recently, a β-l-arabinofuranosidase from the glycoside hydrolase family 127 (HypBA1) was discovered in the newly characterized degradation system of hydroxyproline-linked β-l-arabinooligosaccharides in the bacterium Bifidobacterium longum. Here, we report the crystal structure of HypBA1 in the ligand-free and β-l-arabinofuranose complex forms. The structure of HypBA1 consists of a catalytic barrel domain and two additional β-sandwich domains, with one β-sandwich domain involved in the formation of a dimer. Interestingly, there is an unprecedented metal-binding motif with Zn(2+) coordinated by glutamate and three cysteines in the active site. The glutamate residue is located far from the anomeric carbon of the β-l-arabinofuranose ligand, but one cysteine residue is appropriately located for nucleophilic attack for glycosidic bond cleavage. The residues around the active site are highly conserved among GH127 members. Based on biochemical experiments and quantum mechanical calculations, a possible reaction mechanism involving cysteine as the nucleophile is proposed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G1001
D-Glutamic acid, ≥99% (TLC)
C5H9NO4
A9524
DL-Arabinose, ≥98%
C5H10O5
14495 L-Cysteine 0.1 M solution
G2128
L-Glutamic acid hydrochloride, ≥99% (HPLC)
C5H9NO4 · HCl
209988
Zinc, dust, <10 μm, ≥98%
Zn
324930
Zinc, powder, <150 μm, 99.995% trace metals basis
Zn
243469
Zinc, granular, 20-30 mesh, ACS reagent, ≥99.8%
Zn
31653
Zinc, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.9%, granular
Zn
96454
Zinc, purum, powder
Zn
267619
Zinc, foil, thickness 0.25 mm, 99.999% trace metals basis
Zn
578002
Zinc, nanopowder, 40-60 nm avg. part. size, ≥99% trace metals basis
Zn
356018
Zinc, foil, thickness 0.25 mm, 99.9% trace metals basis
Zn
565148
Zinc, granular, 30-100 mesh, 99%
Zn
267635
Zinc, shot, 5 mm, 99.999% trace metals basis
Zn
267929
Zinc, wire, diam. 1.0 mm, 99.999% trace metals basis
Zn
266361
Zinc, sticks, diam. 7-10 mm, 99.97% trace metals basis
Zn
402583
Zinc, shot, <12 mm, 99.99% trace metals basis
Zn
215481
Zinc, mossy, ≥99%
Zn
266353
Zinc, pieces, 2-14 mesh, 99.9% trace metals basis
Zn
GF86667812
Zinc, foil, 100x100mm, thickness 1.0mm, as rolled, 99.95+%
Zn
716723
Zinc, shot, 99.9999%
Zn
349410
Zinc, foil, thickness 1.0 mm, 99.99% trace metals basis
Zn
215503
Zinc, mossy, 99.99% trace metals basis
Zn
GF60963453
Zinc, wire reel, 0.5m, diameter 0.5mm, as drawn, 99.99+%
Zn
GF63340758
Zinc, foil, 100x100mm, thickness 0.20mm, as rolled, 99.95+%
Zn
749060
Zinc, sputtering target, diam. × thickness 3.00 in. × 0.125 in., 99.995% trace metals basis
Zn
GF59993061
Zinc, foil, light tested, 100x100mm, thickness 0.025mm, as rolled, 99.95+%
Zn
GF37287704
Zinc, wire reel, 100m, diameter 0.25mm, as drawn, 99.99+%
Zn
GF32078853
Zinc, foil, light tested, 150x150mm, thickness 0.05mm, as rolled, 99.95+%
Zn
GF72714102
Zinc, wire reel, 0.1m, diameter 1.0mm, 99.999%
Zn