EMAIL THIS PAGE TO A FRIEND
Biological & pharmaceutical bulletin
Antidepressant activity of astilbin: involvement of monoaminergic neurotransmitters and BDNF signal pathway.
PMID 24681540
Abstract
Depression and related mood disorders are among the world's greatest public health problems. Previous studies have demonstrated that astilbin (AST) has broad pharmacological functions which may modulate numerous pathways, such as antioxidant, scavenging free radicals, anti-inflammatory and so on, similarly to some of other flavonoids. In this study, the antidepressant-like effect of AST was investigated using chronic unpredictable mild stress (CUMS) model of depression in mice. The results showed that chronic administration of AST at doses of 10, 20 and 40 mg/kg (intraperitoneally (i.p.), 21 d) reduced depressive-like behaviors of mice in the forced swim test (FST), tail suspension test (TST) and sucrose preference test (SPT) without affecting locomotor activity. AST increased the contents of serotonin (5-HT) and dopamine (DA) in the frontal cortex of CUMS mice. Additionally, it was shown that AST treatment restored the CUMS-induced inhibition of extracellular signal-regulated kinase (ERK) 1/2 and AKT phosphorylation in the frontal cortex, conformed to the brain-derived neurotrophic factor (BDNF) expression. Our findings suggest that AST has antidepressant activities and the mechanisms, at least in part, relate to up-regulation of monoaminergic neurotransmitters (5-HT and DA) and activation of the BDNF signaling pathway.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4500800
Anti-AKT antibody produced in rabbit, affinity isolated antibody
SAB4300335
Anti-AKT1 (Ab-308) antibody produced in rabbit, affinity isolated antibody
SAB4300575
Anti-AKT1 (Ab-450) antibody produced in rabbit, affinity isolated antibody
GW22757 Anti-AKT1 (ab1) antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB3500216
Anti-Akt1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
AV06008 Anti-AKT1 antibody produced in rabbit, affinity isolated antibody
SAB4300493
Anti-MAPK3 (Ab-202) antibody produced in rabbit, affinity isolated antibody
SAB4300494
Anti-MAPK3 (Ab-204) antibody produced in rabbit, affinity isolated antibody
SAB1412386
ANTI-MAPK3 antibody produced in mouse, clone 1E2, purified immunoglobulin, buffered aqueous solution
SAB2701851
Anti-mapk3 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2701770
Anti-mapk3 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA003995
Anti-MAPK3 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA005700
Anti-MAPK3 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
M3946 Anti-MNK1 antibody produced in rabbit, ~1.4 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB4503422
Anti-MNK1 antibody produced in rabbit, affinity isolated antibody
SAB4503423
Anti-MNK1 antibody produced in rabbit, affinity isolated antibody
SAB4502389
Anti-p44 MAP Kinase antibody produced in rabbit, affinity isolated antibody
SAB4502390
Anti-p44 MAP Kinase antibody produced in rabbit, affinity isolated antibody
WH0000207M3
Monoclonal Anti-AKT1 antibody produced in mouse, clone 2E11, purified immunoglobulin, buffered aqueous solution
SAB1400005
Monoclonal Anti-AKT1 antibody produced in mouse, clone 6D6, purified immunoglobulin, buffered aqueous solution
SAB1400006
Monoclonal Anti-AKT1 antibody produced in mouse, clone 6G6, purified immunoglobulin, buffered aqueous solution
SAB1400007
Monoclonal Anti-AKT1 antibody produced in mouse, clone 2H1, purified immunoglobulin, buffered aqueous solution
SAB1400008
Monoclonal Anti-AKT1 antibody produced in mouse, clone 4B9, purified immunoglobulin, buffered aqueous solution
WH0005595M1
Monoclonal Anti-MAPK3 antibody produced in mouse, clone 3C9, purified immunoglobulin, buffered aqueous solution
SAB5300429
Monoclonal Anti-MAPK3 antibody produced in mouse, clone 1E5, ascites fluid
R2650 Monoclonal Anti-Rac antibody produced in mouse, clone 23A8, purified immunoglobulin, buffered aqueous solution
PLA0238
Rabbit anti-AKT1 Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.
PLA0235
Rabbit anti-ERK1 Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.