EMAIL THIS PAGE TO A FRIEND
Inflammation
Bone morphogenic protein-7 contributes to cerebral ischemic preconditioning induced-ischemic tolerance by activating p38 mitogen-activated protein kinase signaling pathway.
PMID 24682853