EMAIL THIS PAGE TO A FRIEND
Anti-cancer drugs
Superior activity of a new histone deacetylase inhibitor (ZYJ-34c) in inhibiting growth of human leukemia cells by inducing p21WAF1 expression and cell cycle arrest.
PMID 24686006
Abstract
Histone deacetylase inhibitors are a new class of anticancer agents that inhibit cancer cell proliferation and induce apoptosis and cell cycle arrest in various cancer cells. Recently, we identified ZYJ-34c, a modified histone deacetylase inhibitor that showed significantly higher antitumor activity in vivo than the FDA-approved drug suberoylanilide hydroxamic acid. We aimed to investigate its exact mechanism of action. Activity of ZY-34c against human erythroleukemic (K562) cells, human myeloid leukemia (HL-60) cells, and primary leukemic cells were investigated in vitro using proliferation assays, cell cycle assays, apoptosis assays, RNA interference, promoter acetylation assays, and assays of transcription of related molecules. ZYJ-34c strongly inhibited the proliferation of leukemia cells compared with suberoylanilide hydroxamic acid. Primary leukemic cells isolated from patients with acute myeloid leukemia were more sensitive to ZYJ-34c. The 50% growth-inhibitory concentrations of ZYJ-34c and suberoylanilide hydroxamic acid were 2.95±0.75 and 4.45±0.29 μmol/l for K562 cells treated for 48 h, respectively. We found that ZYJ-34c caused more significant G1 cell cycle arrest than suberoylanilide hydroxamic acid, in a time-dependent manner. ZYJ-34c-induced hyperacetylation of histone H3 and H4 around the promoter region of p21. P21 RNA interference markedly impaired ZYJ-34c-induced or suberoylanilide hydroxamic acid-induced G1 cell cycle arrest. In addition, levels of bcr-abl mRNA and protein in K562 cells decreased after treatment with either suberoylanilide hydroxamic acid or ZYJ-34c; moreover, ZYJ-34c had a higher inhibition activity. ZYJ-34c could represent a novel pharmacological agent with potential benefit for patients with leukemia.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
HPA049830
Anti-KRAS antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2107115
Anti-KRAS antibody produced in rabbit, affinity isolated antibody
SAB2101297
Anti-KRAS antibody produced in rabbit, affinity isolated antibody
SAB4100599
Anti-KRAS antibody, Mouse monoclonal, clone KRAS-A582, purified from hybridoma cell culture
SAB2700181
Anti-NS3 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4501190
Anti-NSG1 antibody produced in rabbit, affinity isolated antibody
SAB4503030
Anti-TCTP antibody produced in rabbit, affinity isolated antibody
R3400 Monoclonal Anti-c-K-Ras antibody produced in mouse, clone 234-4.2, purified immunoglobulin, buffered aqueous solution
WH0027065M1
Monoclonal Anti-D4S234E antibody produced in mouse, clone 1C3, purified immunoglobulin, buffered aqueous solution
WH0003845M1
Monoclonal Anti-KRAS antibody produced in mouse, clone 3B10-2F2, purified immunoglobulin, buffered aqueous solution
SAB1404011
Monoclonal Anti-KRAS antibody produced in mouse, clone 4F3, purified immunoglobulin, buffered aqueous solution
WH0007178M1
Monoclonal Anti-TPT1 antibody produced in mouse, clone 3C7, purified immunoglobulin, buffered aqueous solution
SAB2105613
Anti-D4S234E, (N-terminal) antibody produced in rabbit, affinity isolated antibody
SAB2502090
Anti-Nsg1 (N-terminal) antibody produced in goat, affinity isolated antibody, buffered aqueous solution