EMAIL THIS PAGE TO A FRIEND
Japanese journal of clinical oncology
Avoidance of false-positive findings on ¹⁸F-FDG-PET/CT using PET/ultrasound fusion: displaced laryngeal silicone implant versus recurrent cancer.
PMID 24691172