EMAIL THIS PAGE TO A FRIEND
The Journal of Clinical Endocrinology and Metabolism
Associations between insulin resistance, free fatty acids, and oocyte quality in polycystic ovary syndrome during in vitro fertilization.
PMID 24694334