EMAIL THIS PAGE TO A FRIEND
Journal of Pharmaceutical and Biomedical Analysis
Chemical comparison of Tripterygium wilfordii and Tripterygium hypoglaucum based on quantitative analysis and chemometrics methods.
PMID 24694566