EMAIL THIS PAGE TO A FRIEND
Therapeutic Drug Monitoring
Measurement of the direct oral anticoagulants apixaban, dabigatran, edoxaban, and rivaroxaban in human plasma using turbulent flow liquid chromatography with high-resolution mass spectrometry.
PMID 24695356