EMAIL THIS PAGE TO A FRIEND
The Journal of Pharmacy and Pharmacology
Ex-vivo permeation study of chlorin e6-polyvinylpyrrolidone complexes through the chick chorioallantoic membrane model.
PMID 24697155