EMAIL THIS PAGE TO A FRIEND
Insect biochemistry and molecular biology
Highly efficient multiplex targeted mutagenesis and genomic structure variation in Bombyx mori cells using CRISPR/Cas9.
PMID 24698835
Abstract
Bombyx mori is an economically important insect and a model organism for studying lepidopteran and arthropod biology. Using a highly efficient CRISPR/Cas9 system, we showed that this system could mediate highly efficient targeted genome editing of a single gene locus, large chromosomal deletion or inversion, and also multiplex genome editing of 6 genes simultaneously in BmNs cell line derived from B. mori. The simplicity and high efficiency of our system provide unprecedented possibilities for researchers to implement precise and sophisticated manipulation of a chosen B. mori gene in BmNs cells easily in a limited time course, and perhaps new opportunities for functional genomics of B. mori and other lepidopteran insects.