EMAIL THIS PAGE TO A FRIEND
The American journal of gastroenterology
Rifaximin for episodic, overt hepatic encephalopathy: the data are catching up to clinical practice, but questions remain.
PMID 24698864
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Y0001510
Lactulose for peak identification, European Pharmacopoeia (EP) Reference Standard
C12H22O11