EMAIL THIS PAGE TO A FRIEND
Nature structural & molecular biology
New insights into ubiquitin E3 ligase mechanism.
PMID 24699078
Abstract
E3 ligases carry out the final step in the ubiquitination cascade, catalyzing transfer of ubiquitin from an E2 enzyme to form a covalent bond with a substrate lysine. Three distinct classes of E3 ligases have been identified that stimulate transfer of ubiquitin and ubiquitin-like proteins through either a direct or an indirect mechanism. Only recently have the catalytic mechanisms of E3 ligases begun to be elucidated.