EMAIL THIS PAGE TO A FRIEND
Nature structural & molecular biology
Lysine-targeting specificity in ubiquitin and ubiquitin-like modification pathways.
PMID 24699079
Abstract
Ubiquitin and ubiquitin-like modifications are central to virtually all cellular signaling pathways. They occur primarily on lysine residues of target proteins and stimulate a large number of downstream signals. The diversity of these signals depends on the type, location and dynamics of the modification, but the role of the exact site of modification and the selectivity for specific lysines are poorly understood. Here we review the current literature on lysine specificity in these modifications, and we highlight the known signaling mechanisms and the open questions that pose future challenges to ubiquitin research.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Y0000397
Lysine acetate, European Pharmacopoeia (EP) Reference Standard
C6H14N2O2 · C2H4O2