EMAIL THIS PAGE TO A FRIEND
Environmental toxicology and pharmacology
Cytotoxic effects of three new metabolites from Red Sea marine sponge, Petrosia sp.
PMID 24699241
Abstract
Marine sponges represent an affluent source of biogenetically unprecedented array of biologically active compounds. This study revealed the isolation of ten compounds from marine sponge of Petrosia sp. Their chemical structures were determined by using 1D and 2D NMR, UV, IR and MS measurements. A polyoxygenated steroid (3β,7β,9α-trihydroxycholest-5-en (1), a purine-derivative (3,7-dimethyl-2-(methylamino)-3H-purin-6(7H)-one (2) and a sphingolipid (N-((3S,E)-1,3-dihydroxytetracos-4-en-2-yl)stearamide (3) proved to be new compounds. Meanwhile, seven known compounds; (4-10) were also identified. The cytotoxicity of the total extract and the isolated compounds were subjected to cytotoxicity evaluation employing two cancer cell lines; HepG2 and MCF-7. All tested compounds exhibited cytotoxic effect on both cancer cell lines with IC(50) in range of 20-500 μM. The proposed mechanism of cytotoxic activities was examined through its molecular affinity to the DNA. Compound 5 showed the highest affinity to the DNA with IC(50) 30 μg/mL.