EMAIL THIS PAGE TO A FRIEND
Oncology reports
SOX2 promotes the migration and invasion of laryngeal cancer cells by induction of MMP-2 via the PI3K/Akt/mTOR pathway.
PMID 24700142
Abstract
SOX2 is a high mobility group box containing transcription factor that has been reported to be aberrantly overexpressed in various human malignancies, including laryngeal squamous cell carcinoma (LSCC). However, the potential role of SOX2 in LSCC migration and invasion remains to be elucidated. In the present study, we generated stable transformants of human LSCC cells constitutively overexpressing SOX2 and investigated the effects of SOX2 overexpression on migration and invasion in LSCC cells as well as the possible underlying mechanisms. We found that ectopic overexpression of SOX2 in LSCC cells enhanced their migratory and invasive ability in vitro, accompanied by increased expression and activity of matrix metalloproteinase (MMP)-2. Meanwhile, SOX2-induced cell migration and invasion were significantly abrogated by a neutralizing anti-MMP-2 antibody or small interfering RNA targeting MMP-2. Furthermore, overexpression of SOX2 induced phosphorylation of Akt and mammalian target of rapamycin (mTOR), which are downstream effectors of the PI3K pathway. Finally, LY294002, an inhibitor of PI3K, also markedly abolished SOX2-induced activation of the Akt/mTOR pathway and increased cell invasion and MMP-2 expression. Taken together, we conclude that SOX2 promotes migration and invasion of laryngeal cancer cells by inducing MMP-2 via the PI3K/Akt/mTOR pathway. Our findings suggest that SOX2 may serve as a potential therapeutic target for LSCC.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4500799
Anti-AKT antibody produced in rabbit, affinity isolated antibody
SAB4500800
Anti-AKT antibody produced in rabbit, affinity isolated antibody
SAB4500802
Anti-AKT antibody produced in rabbit, affinity isolated antibody
SAB4300335
Anti-AKT1 (Ab-308) antibody produced in rabbit, affinity isolated antibody
SAB4300575
Anti-AKT1 (Ab-450) antibody produced in rabbit, affinity isolated antibody
SAB4300334
Anti-AKT1 (Ab-473) antibody produced in rabbit, affinity isolated antibody
GW22757 Anti-AKT1 (ab1) antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB3500216
Anti-Akt1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4500007
Anti-AKT1 antibody produced in rabbit, affinity isolated antibody
AV06008 Anti-AKT1 antibody produced in rabbit, affinity isolated antibody
HPA002891
Anti-AKT1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2100362
Anti-CCND1 (ab1) antibody produced in rabbit, affinity isolated antibody
SAB2100363
Anti-CCND1 (ab2) antibody produced in rabbit, affinity isolated antibody
SAB2100364
Anti-CCND1 (ab3) antibody produced in rabbit, affinity isolated antibody
SAB1405510
Anti-CCND1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
M4065 Anti-Matrix Metalloproteinase-2 antibody produced in rabbit, ~1 mg/mL, IgG fraction of antiserum, buffered aqueous solution
HPA001939
Anti-MMP2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2502098
Anti-PCNA antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB4502103
Anti-PCNA antibody produced in rabbit, affinity isolated antibody
HPA030521
Anti-PCNA antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab1
HPA030522
Anti-PCNA antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab2
HPA030523
Anti-PCNA antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab3
SAB2701819
Anti-PCNA antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4503853
Anti-phospho-Akt (pSer124) antibody produced in rabbit, affinity isolated antibody
SAB4504016
Anti-phospho-Akt (pSer129) antibody produced in rabbit, affinity isolated antibody
SAB4504331
Anti-phospho-Akt (pSer473) antibody produced in rabbit, affinity isolated antibody
SAB4504332
Anti-phospho-Akt (pThr308) antibody produced in rabbit, affinity isolated antibody
SAB4504017
Anti-phospho-Akt (pThr72) antibody produced in rabbit, affinity isolated antibody
SAB4504110
Anti-phospho-Akt (pTyr326) antibody produced in rabbit, affinity isolated antibody
SAB4504014
Anti-phospho-Akt (pTyr474) antibody produced in rabbit, affinity isolated antibody