EMAIL THIS PAGE TO A FRIEND
Journal of Pharmaceutical Sciences
Soy phosphatidylinositol containing nanoparticle prolongs hemostatic activity of B-domain deleted factor VIII in hemophilia A mice.
PMID 24700333