EMAIL THIS PAGE TO A FRIEND
Journal of Pharmaceutical Sciences
Small-angle x-ray scattering screening complements conventional biophysical analysis: comparative structural and biophysical analysis of monoclonal antibodies IgG1, IgG2, and IgG4.
PMID 24700358