EMAIL THIS PAGE TO A FRIEND
The Medical journal of Australia
Staphylococcus aureus bacteraemia associated with peripherally inserted central catheters: the role of chlorhexidine gluconate-impregnated sponge dressings.
PMID 24702084