EMAIL THIS PAGE TO A FRIEND
Journal of Clinical Pharmacy and Therapeutics
Genetic selection of volunteers and concomitant dose adjustment leads to comparable hydralazine/valproate exposure.
PMID 24702251