EMAIL THIS PAGE TO A FRIEND
The Journal of urology
Re: Renal ammonium excretion after an acute acid load: blunted response in uric acid stone formers but not in patients with type 2 diabetes.
PMID 24703125