EMAIL THIS PAGE TO A FRIEND
Chemico-Biological Interactions
Artesunate induces apoptosis through caspase-dependent and -independent mitochondrial pathways in human myelodysplastic syndrome SKM-1 cells.
PMID 24704559
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A0835
N-Acetyl-Asp-Glu-Val-Asp-al, ≥95%, powder
C20H30N4O11