EMAIL THIS PAGE TO A FRIEND
The British journal of nutrition
Effect of tryptophan supplementation on diet-induced non-alcoholic fatty liver disease in mice.
PMID 24708895
Abstract
Intestinal serotonin (5-hydroxytrypamine, 5-HT) metabolism is thought to play a role in gut functions by regulating motility, permeability and other functions of the intestine. In the present study, we investigated the effect of tryptophan (TRP), the precursor of 5-HT, supplementation on intestinal barrier functions and non-alcoholic fatty liver disease (NAFLD). An established mouse model of NAFLD induced by feeding a fructose-rich diet (N group) was used in the present study. TRP was administered orally for 8 weeks to C57BL/6J control or NAFLD mice. NAFLD-related liver parameters (hepatic TAG and Oil Red O staining), intestinal barrier parameters (tight-junction protein occludin and portal plasma lipopolysaccharides (LPS)) and 5-HT-related parameters (5-HT, 5-HT transporter (SERT) and motility) were measured. We observed reduced duodenal occludin protein concentrations (P= 0·0007), high portal plasma LPS concentrations (P= 0·005) and an elevated liver weight:body weight ratio (P= 0·01) in the N group compared with the parameters in the control group. TRP supplementation led to an increase in occludin concentrations (P= 0·0009) and consecutively reduced liver weight:body weight ratio (P= 0·009) as well as overall hepatic fat accumulation in the N group (P= 0·05). In addition, the N group exhibited reduced SERT protein expression (P= 0·002), which was normalised by TRP supplementation (P= 0·02). For the first time, our data indicate that oral TRP supplementation attenuates experimental NAFLD in mice. The underlying mechanisms are not clear, but probably involve stabilisation of the intestinal barrier in the upper small intestine and amelioration of the dysregulated intestinal serotonergic system.