EMAIL THIS PAGE TO A FRIEND
International Journal of Pharmaceutics
An improved method for the characterization of supersaturation and precipitation of poorly soluble drugs using pulsatile microdialysis (PMD).
PMID 24709210