EMAIL THIS PAGE TO A FRIEND
American journal of kidney diseases : the official journal of the National Kidney Foundation
Membranous nephropathy with crescents: a series of 19 cases.
PMID 24709471
Abstract
Membranous nephropathy (MN) with crescents is rare and, in the absence of lupus, usually is associated with anti-glomerular basement membrane (anti-GBM) nephritis or antineutrophil cytoplasmic antibody (ANCA)-positive glomerulonephritis. Only rare cases of crescentic MN without ANCA or anti-GBM have been reported. Case series. 19 patients with ANCA- and anti-GBM-negative crescentic MN and no clinical evidence of systemic lupus. Clinical features, kidney biopsy findings, laboratory results, treatment, and follow-up of patients with crescentic MN. Mean age was 55 (range, 5-86) years. All patients presented with proteinuria (mean protein excretion, 11.5 [range, 3.3-29] g/d) and nearly all had hematuria; 16 of 19 (84%) patients had decreased estimated glomerular filtration rates (eGFRs; mean serum creatinine, 2.9 [range, 0.4-10] mg/dL; mean eGFR, 39.7 [range, 4 to >100] mL/min/1.73 m2). Glomeruli showed on average 25% (range, 2%-73%) involvement by crescents. All showed a membranous pattern; 7 showed mesangial and 2 showed segmental endocapillary proliferation. By immunofluorescence, all cases showed granular subepithelial immunoglobulin G (IgG) and κ and λ light chains, and all but one showed C3; 5 showed C1q or IgA. Electron microscopy revealed stages I-III MN; 38% of cases were M-type phospholipase A2 receptor (PLA2R) associated, indicating that at least some were primary MN. Follow-up clinical data were available for all patients (mean, 22 [range, 1.5-138] months). 14 patients received immunosuppressive therapy, and 2, only angiotensin-converting enzyme inhibitor/angiotensin receptor blocker therapy. 4 patients (21%) progressed to end-stage renal disease, at 0-9 months postbiopsy. Mean serum creatinine level of those without end-stage renal disease at follow-up was 1.7 (range, 0.5-4.1) mg/dL; mean eGFR was 53.3 (range, 16-103) mL/min/1.73 m2. 67% of patients had proteinuria with protein excretion≥1 (mean, 3.2) g/d at follow-up. Retrospective study. Crescentic MN is a rare variant of MN that usually presents with heavy proteinuria, hematuria, and decline in GFR. The prognosis is variable and the disease may respond to therapy, but most patients develop a long-term decline in GFR.
Related Materials