EMAIL THIS PAGE TO A FRIEND
Cancer research
HMMR maintains the stemness and tumorigenicity of glioblastoma stem-like cells.
PMID 24710409
Abstract
Glioblastoma (GBM) stem cells (GSC) are a subpopulation of tumor cells that display stem-like characteristics (stemness) and play unique roles in tumor propagation, therapeutic resistance, and tumor recurrence. Therapeutic targets in GSCs are a focus of increasing interest to improve GBM therapy. Here we report that the hyaluronan-mediated motility receptor (HMMR) is highly expressed in GBM tumors, where it supports the self-renewal and tumorigenic potential of GSCs. HMMR silencing impairs GSC self-renewal and inhibits the expression of GSC markers and regulators. Furthermore, HMMR silencing suppresses GSC-derived tumor growth and extends the survival of mice bearing GSC xenografts. Conversely, HMMR overexpression promotes GSC self-renewal and intracranial tumor propagation. In human GBM tumor specimens, HMMR expression is correlated positively with the expression of stemness-associated markers and regulators. Our findings identify HMMR as a candidate therapeutic target to GSCs as a GBM treatment strategy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4200033
Anti-BMI1 antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution
HPA030472
Anti-BMI1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2106309
Anti-BMI1 antibody produced in rabbit, affinity isolated antibody
SAB4300493
Anti-MAPK3 (Ab-202) antibody produced in rabbit, affinity isolated antibody
SAB4300494
Anti-MAPK3 (Ab-204) antibody produced in rabbit, affinity isolated antibody
GW22781 Anti-MAPK3 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB1412386
ANTI-MAPK3 antibody produced in mouse, clone 1E2, purified immunoglobulin, buffered aqueous solution
SAB2701851
Anti-mapk3 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2701770
Anti-mapk3 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA003995
Anti-MAPK3 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA005700
Anti-MAPK3 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2104574
Anti-MAPK3 antibody produced in rabbit, affinity isolated antibody
SAB2104732
Anti-MAPK3 antibody produced in rabbit, affinity isolated antibody
M3946 Anti-MNK1 antibody produced in rabbit, ~1.4 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB4503422
Anti-MNK1 antibody produced in rabbit, affinity isolated antibody
SAB4503423
Anti-MNK1 antibody produced in rabbit, affinity isolated antibody
SAB4502389
Anti-p44 MAP Kinase antibody produced in rabbit, affinity isolated antibody
SAB4502390
Anti-p44 MAP Kinase antibody produced in rabbit, affinity isolated antibody
AV34213
Anti-PCGF4 antibody produced in rabbit, affinity isolated antibody
SAB4300132
Anti-phospho-MAP2K1 (pSer221) antibody produced in rabbit, affinity isolated antibody
SAB4300234
Anti-phospho-MAP2K1 (pThr291) antibody produced in rabbit, affinity isolated antibody
SAB4504620
Anti-phospho-MAP2K1 (pThr386) antibody produced in rabbit, affinity isolated antibody
SAB4300194
Anti-phospho-MAPK3 (pThr202) antibody produced in rabbit, affinity isolated antibody
SAB4300195
Anti-phospho-MAPK3 (pTyr204) antibody produced in rabbit, affinity isolated antibody
SAB4504038
Anti-phospho-MEK1 (pSer298) antibody produced in rabbit, affinity isolated antibody
SAB4504039
Anti-phospho-MEK1 (pThr286) antibody produced in rabbit, affinity isolated antibody
SAB4504468
Anti-phospho-MEK1 (pThr291) antibody produced in rabbit, affinity isolated antibody
SAB1306660 ANTI-PHOSPHO-MEK1(S222) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4504626
Anti-phospho-MNK1 (pThr255) antibody produced in rabbit, affinity isolated antibody
SAB4504714
Anti-phospho-Mnk1 (pThr385) antibody produced in rabbit, affinity isolated antibody