EMAIL THIS PAGE TO A FRIEND
Dermatologic surgery : official publication for American Society for Dermatologic Surgery [et al.]
Prevention of thyroidectomy scars in Korean patients using a new combination of intralesional injection of low-dose steroid and pulsed dye laser starting within 4 weeks of suture removal.
PMID 24712703
Abstract
Regulation of inflammation during the wound healing process reduces scar formation at the injury site. To evaluate the effect of intralesional injection of low-dose steroid with pulsed dye laser on healing of early postoperative thyroidectomy scars. Twenty Korean women with thyroidectomy scars were enrolled. All were treated with an intralesional injection of low-dose steroid (2 mg/mL) and 595-nm pulsed dye laser starting within 4 weeks of suture removal. The Vancouver Scar Scale (VSS), Global Assessment Score (GAS), and Patient Satisfaction Score were used in this evaluation. Average VSS scores were significantly lower after treatment. The GAS also indicated better cosmetic outcomes after steroid injection in the laser treatment group than after laser treatment only. Early postoperative intralesional injection of low-dose steroid and pulsed dye laser treatment is effective and safe.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T1899990
Triamcinolone, European Pharmacopoeia (EP) Reference Standard
C21H27FO6