Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research
Topological mapping of BRIL reveals a type II orientation and effects of osteogenesis imperfecta mutations on its cellular destination.
PMID 24715519
BRIL/IFITM5 is a membrane protein present almost exclusively in osteoblasts, which is believed to adopt a type III (N-out/C-out) topology. Mutations in IFITM5 cause OI type V, but the characteristics of the mutant protein and the mechanism involved are still unknown. The purpose of the current study was to re-assess the topology, localization, and biochemical properties of BRIL and compare it to the OI type V mutant in MC3T3 osteoblasts. Immunofluorescence labeling was performed with antibodies directed against BRIL N- or C-terminus. In intact cells, BRIL labeling was conspicuously detected at the plasma membrane only with the anti-C antibody. Detection of BRIL N-terminus was only possible after cell permeabilization, revealing both plasma membrane and Golgi labeling. Trypsinization of live cells expressing BRIL only cleaved off the C-terminus, confirming that it is a type II protein and that its N-terminus is intracellular. A truncated form of BRIL lacking the last 18 residues did not appear to affect localization, whereas mutation of a single leucine to arginine within the transmembrane segment abolished plasma membrane targeting. BRIL is first targeted to the endoplasmic reticulum as the entry point to the secretory pathway and rapidly traffics to the Golgi via a COPII-dependent pathway. BRIL was found to be palmitoylated and two conserved cysteine residues (C52 and C53) were critical for targeting to the plasma membrane. The OI type V mutant BRIL, having a five residue extension (MALEP) at its N-terminus, presented with exactly the same topological and biochemical characteristics as wild type BRIL. In contrast, the S42 > L mutant BRIL was trapped intracellularly in the Golgi. BRIL proteins and transcripts were equally detected in bone from a patient with OI type V, suggesting that the cause of the disease is a gain of function mediated by a faulty intracellular activity of the mutant BRIL.