EMAIL THIS PAGE TO A FRIEND
Chemistry (Weinheim An Der Bergstrasse, Germany)
Anti-cancer applications of titanocene-functionalised nanostructured systems: an insight into cell death mechanisms.
PMID 24715574