EMAIL THIS PAGE TO A FRIEND
Veterinaria italiana
Molecular typing of Salmonella enterica subspecies enterica serovar Typhimurium isolated in Abruzzo region (Italy) from 2008 to 2010.
PMID 24715591
Abstract
In this study, 47 antibiotic-resistant strains of Salmonella enterica subspecies enterica serovar Typhimurium (ST) were characterised, including 15 monophasic variants 1, 4, [5], 12:i:-, (STm) isolated from different matrices. They were all selected from 389 Salmonella enterica subspecies enterica strains isolated during 2008-2010 in Abruzzo region (Italy). Thirty-seven strains showed to be resistant to more than 1 antibiotic. Among 47 isolates, phage type U311 and DT104 were identified. The ASSuT resistance pattern was predominant in mST strains and ACSSuT in ST DT104 and U302. A multiplex Polimerase Chain Reaction (PCR) method was used to investigate 4 genes: fluorfenicol (floSt), virulence (spvC), invasine (invA) and integrase (int). All ST the strain were positive for invA gene and 28,32% of strains were positive for spvC gene. PFGE analysis revealed a large number of small clonal populations, however not ascrivable to outbreaks.