EMAIL THIS PAGE TO A FRIEND
The Biochemical Journal
The glycation site specificity of human serum transferrin is a determinant for transferrin's functional impairment under elevated glycaemic conditions.
PMID 24716439