EMAIL THIS PAGE TO A FRIEND
The Journal of neuroscience : the official journal of the Society for Neuroscience
Ionotropic NMDA receptor signaling is required for the induction of long-term depression in the mouse hippocampal CA1 region.
PMID 24719106
Abstract
Previous studies have provided strong support for the notion that NMDAR-mediated increases in postsynaptic Ca(2+) have a crucial role in the induction of long-term depression (LTD). This view has recently been challenged, however, by findings suggesting that LTD induction is instead attributable to an ion channel-independent, metabotropic form of NMDAR signaling. Thus, to explore the role of ionotropic versus metabotropic NMDAR signaling in LTD, we examined the effects of varying extracellular Ca(2+) levels or blocking NMDAR channel ion fluxes with MK-801 on LTD and NMDAR signaling in the mouse hippocampal CA1 region. We find that the induction of LTD in the adult hippocampus is highly sensitive to extracellular Ca(2+) levels and that MK-801 blocks NMDAR-dependent LTD in the hippocampus of both adult and immature mice. Moreover, MK-801 inhibits NMDAR-mediated activation of p38-MAPK and dephosphorylation of AMPAR GluA1 subunits at sites implicated in LTD. Thus, our results indicate that the induction of LTD in the hippocampal CA1 region is dependent on ionotropic, rather than metabotropic, NMDAR signaling.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M107
(+)-MK-801 hydrogen maleate, powder
C16H15N · C4H4O4