EMAIL THIS PAGE TO A FRIEND
Neurology
Autosomal recessive cerebellar ataxia of adult onset due to STUB1 mutations.
PMID 24719489
Abstract
Autosomal recessive ataxias affect about 1 person in 20,000. Friedreich ataxia accounts for one-third of the cases in Caucasians; the others are due to a growing list of very rare molecular defects, including mild forms of metabolic diseases. In nearly 50%, the genetic cause remains undetermined.
Related Materials