EMAIL THIS PAGE TO A FRIEND
Lakartidningen
[Children's iron needs and how we best can protect the child's brain].
PMID 24720019
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C3518
Carbonyl iron, ≥97% Fe basis
Fe
12310
Iron, ≥99%, reduced, powder (fine)
Fe
209309
Iron, powder, −325 mesh, 97%
Fe
255637
Iron, powder, ≥99.99% trace metals basis
Fe
44890
Iron, puriss. p.a., carbonyl-Iron powder, low in magnesium and manganese compounds, ≥99.5% (RT)
Fe
413054
Iron, granular, 10-40 mesh, >99.99% trace metals basis
Fe
267945
Iron, chips, 99.98% trace metals basis
Fe
746843
Iron, nanopowder, 35-45 nm particle size, 99.5% trace metals basis
Fe
338141
Iron, foil, thickness 0.25 mm, ≥99.99% trace metals basis
Fe
356808
Iron, foil, thickness 0.1 mm, ≥99.9% trace metals basis
Fe
356824
Iron, wire, diam. 1.0 mm, ≥99.9% trace metals basis
Fe
266213
Iron, rod, diam. 6.3 mm, 99.98% trace metals basis
Fe
746827
Iron, carbon coated magnetic, nanopowder, 25 nm avg. part. size, 99.5% trace metals basis
Fe
356832
Iron, wire, diam. 0.5 mm, ≥99.9% trace metals basis
Fe
GF13964575
Iron, foil, 50x50mm, thickness 0.25mm, hard, 99.5%
Fe
GF61709799
Iron, foil, 150x150mm, thickness 0.1mm, hard, 99.5%
Fe
GF13723758
Iron, foil, 300x300mm, thickness 0.1mm, hard, 99.5%
Fe
GF86771953
Iron, foil, 1m coil, thickness 0.38mm, hard, 99.5%
Fe
GF58809965
Iron, wire reel, 50m, diameter 0.065mm, hard, 99.5%
Fe
GF97671415
Iron, rod, 500mm, diameter 4.8mm, as drawn, 98+%
Fe
GF70739578
Iron, foil, 10mm disks, thickness 0.01mm, 99.85%
Fe
GF91071648
Iron, foil, 10mm disks, thickness 0.01mm, 99.99+%
Fe
GF93750419
Iron, foil, 10mm disks, thickness 0.020mm, 99.85%
Fe
GF38893315
Iron, foil, 10mm disks, thickness 0.025mm, as rolled, 99.99+%
Fe
GF49755291
Iron, foil, 10mm disks, thickness 0.025mm, hard, 99.5%
Fe
GF17553663
Iron, foil, 10mm disks, thickness 0.038mm, hard, 99.5%
Fe
GF28613235
Iron, foil, 10mm disks, thickness 0.05mm, as rolled, 99.99+%
Fe
GF58999471
Iron, foil, 10mm disks, thickness 0.05mm, hard, 99.5%
Fe
GF04554963
Iron, foil, 10mm disks, thickness 0.075mm, as rolled, 99.99+%
Fe
GF99203991
Iron, foil, 10mm disks, thickness 0.075mm, hard, 99.5%
Fe