EMAIL THIS PAGE TO A FRIEND
Skinmed
Disseminated extragenital lichen sclerosus et atrophicus treated with acitretin.
PMID 24720089
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
1011018 Acitretin Related Compound A, United States Pharmacopeia (USP) Reference Standard
C21H26O3