EMAIL THIS PAGE TO A FRIEND
Biomacromolecules
PEG-peptide conjugates.
PMID 24720400
Abstract
The remarkable diversity of the self-assembly behavior of PEG-peptides is reviewed, including self-assemblies formed by PEG-peptides with β-sheet and α-helical (coiled-coil) peptide sequences. The modes of self-assembly in solution and in the solid state are discussed. Additionally, applications in bionanotechnology and synthetic materials science are summarized.