EMAIL THIS PAGE TO A FRIEND
European journal of immunology
IFN-γ-producing NKT cells exacerbate sepsis by enhancing C5a generation via IL-10-mediated inhibition of CD55 expression on neutrophils.
PMID 24723363
Abstract
A role for NKT cells has been implicated in sepsis, but the mechanism by which NKT cells contribute to sepsis remains unclear. Here, we examined WT and NKT-cell-deficient mice of C57BL/6 background during cecal ligation and puncture-induced sepsis. The levels of C5a, IFN-γ, and IL-10 were higher in the serum and peritoneal fluid of WT mice than in those of CD1d(-/-) mice, while the mortality rate was lower in CD1d(-/-) mice than in WT mice. C5a blockade decreased mortality of WT mice during sepsis, whereas it did not alter that of CD1d(-/-) mice. As assessed by intracellular staining, NKT cells expressed IFN-γ, while neutrophils expressed IL-10. Upon coculture, IL-10-deficient NKT cells enhanced IL-10 production by WT, but not IFN-γR-deficient, neutrophils. Meanwhile, CD1d(-/-) mice exhibited high CD55 expression on neutrophils during sepsis, whereas those cells from WT mice expressed minimal levels of CD55. Recombinant IL-10 administration into CD1d(-/-) mice reduced CD55 expression on neutrophils. Furthermore, adoptive transfer of sorted WT, but not IFN-γ-deficient, NKT cells into CD1d(-/-) mice suppressed CD55 expression on neutrophils, but increased IL-10 and C5a levels. Taken together, IFN-γ-producing NKT cells enhance C5a generation via IL-10-mediated inhibition of CD55 expression on neutrophils, thereby exacerbating sepsis.