EMAIL THIS PAGE TO A FRIEND
Neuron
Roles of heat shock factor 1 in neuronal response to fetal environmental risks and its relevance to brain disorders.
PMID 24726381
Abstract
Prenatal exposure of the developing brain to various environmental challenges increases susceptibility to late onset of neuropsychiatric dysfunction; still, the underlying mechanisms remain obscure. Here we show that exposure of embryos to a variety of environmental factors such as alcohol, methylmercury, and maternal seizure activates HSF1 in cerebral cortical cells. Furthermore, Hsf1 deficiency in the mouse cortex exposed in utero to subthreshold levels of these challenges causes structural abnormalities and increases seizure susceptibility after birth. In addition, we found that human neural progenitor cells differentiated from induced pluripotent stem cells derived from schizophrenia patients show higher variability in the levels of HSF1 activation induced by environmental challenges compared to controls. We propose that HSF1 plays a crucial role in the response of brain cells to prenatal environmental insults and may be a key component in the pathogenesis of late-onset neuropsychiatric disorders.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
24102
Ethanol, absolute, ≥99.8% (GC)
C2H6O
24105
Ethanol, puriss., meets analytical specification of Ph. Eur., BP, 96.0-97.2%
C2H6O
24194
Ethanol, absolute, denaturated with 0.5-1.5 Vol.% 2-butanone and approx. 0.001% Bitrex (GC), ≥98% (GC)
C2H6O
E-034
Ethanol Calibration Kit, ampule of 10 × 1.2 mL, certified reference material
C2H6O
E-040
Ethanol-10, 10 mg/dL in H2O, pack of 10 × 1.2 mL ampules, certified reference material
C2H6O
E-031
Ethanol-100 (10 ampules/kit), 100 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material
C2H6O
E-038
Ethanol-100 (5 ampules/kit), 100 mg/dL in H2O, ampule of 5 × 5 mL, certified reference material
C2H6O
E-041
Ethanol-150, 150 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material
C2H6O
E-056
Ethanol-20 (10 ampules/kit), 20 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material
C2H6O
E-043
Ethanol-20 (5 ampules/kit), 20 mg/dL in H2O, ampule of 5 × 5 mL, certified reference material
C2H6O
E-032
Ethanol-200 (10 ampules/kit), 200 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material
C2H6O
E-039
Ethanol-200 (5 ampules/kit), 200 mg/dL in H2O, ampule of 5 × 5 mL, certified reference material
C2H6O
E-035
Ethanol-25, 25 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material
C2H6O
E-033
Ethanol-300, 300 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material
C2H6O
E-045
Ethanol-40, 40 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material
C2H6O
E-036
Ethanol-400 (10 ampules/kit), 400 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material
C2H6O
E-044
Ethanol-400 (5 ampules/kit), 400 mg/dL in H2O, ampule of 5 × 5 mL, certified reference material
C2H6O
E-029
Ethanol-50, 50 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material
C2H6O
E-053
Ethanol-500, 500 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material
C2H6O
E-030
Ethanol-80, 80 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material
C2H6O
E-037
Ethanol-80, 80 mg/dL in H2O, ampule of 5 × 5 mL, certified reference material
C2H6O
187380 Ethyl alcohol, denatured, reagent grade
459844
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
C2H6O
E7023
Ethyl alcohol, Pure, 200 proof, for molecular biology
C2H6O
493546
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
C2H6O
459828
Ethyl alcohol, Pure, 200 proof, HPLC/spectrophotometric grade
C2H6O
E7148
Ethyl alcohol, Pure, 190 proof, for molecular biology
C2H6O
270741
Reagent Alcohol, for HPLC
277649
Reagent Alcohol, anhydrous, ≤0.003% water
676829
Reagent Alcohol, anhydrous, ≤0.005% water