EMAIL THIS PAGE TO A FRIEND
Drug and Alcohol Dependence
Sex differences in methamphetamine pharmacokinetics in adult rats and its transfer to pups through the placental membrane and breast milk.
PMID 24726427