EMAIL THIS PAGE TO A FRIEND
Biochemical pharmacology
Isoquercitrin and polyphosphate co-enhance mineralization of human osteoblast-like SaOS-2 cells via separate activation of two RUNX2 cofactors AFT6 and Ets1.
PMID 24726443
Abstract
Isoquercitrin, a dietary phytoestrogen, is a potential stimulator of bone mineralization used for prophylaxis of osteoporotic disorders. Here we studied the combined effects of isoquercitrin, a cell membrane permeable 3-O-glucoside of quercetin, and polyphosphate [polyP], a naturally occurring inorganic polymer inducing bone formation, on mineralization of human osteoblast-like SaOS-2 cells. Both compounds isoquercitrin and polyP induce at non-toxic concentrations the mineralization process of SaOS-2 cells. Co-incubation experiments revealed that isoquercitrin (at 0.1 and 0.3μM), if given simultaneously with polyP (as Ca(2+) salt; at 3, 10, 30 and 100μM) amplifies the mineralization-enhancing effect of the inorganic polymer. The biomineralization process induced by isoquercitrin and polyP is based on two different modes of action. After incubation of the cells with isoquercitrin or polyP the expression of the Runt-related transcription factor 2 [RUNX2] is significantly upregulated. In addition, isoquercitrin causes a strong increase of the steady-state-levels of the two co-activators of RUNX2, the activating transcription factor 6 [ATF6] and the Ets oncogene homolog 1 [Ets1]. The activating effect of isoquercitrin occurs via a signal transduction pathway involving ATF6, and by that, is independent from the induction cascade initiated by polyP. This conclusion is supported by the finding that isoquercitrin upregulates the expression of the gene encoding for osteocalcin, while polyP strongly increases the expression of the Ets1 gene and of the alkaline phosphatase. We show that the two compounds, polyP and isoquercitrin, have a co-enhancing effect on bone mineral formation and in turn might be of potential therapeutic value for prevention/treatment of osteoporosis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Y0001568 Isoquercitroside, European Pharmacopoeia (EP) Reference Standard
C21H20O12