EMAIL THIS PAGE TO A FRIEND
Experimental parasitology
Combined drug therapy in the management of granulomatous amoebic encephalitis due to Acanthamoeba spp., and Balamuthia mandrillaris.
PMID 24726699
Abstract
Granulomatous amoebic encephalitis (GAE) is caused by two protist pathogens, Acanthamoeba spp., and Balamuthia mandrillaris. Although rare, it almost always results in death. In the present study, amoebae were treated with various combinations of clinically-approved drugs, targeting vital cellular receptors and biochemical pathways. The results revealed that among the seven different combinations tested, three proved highly effective against both Acanthamoeba castellanii as well as B. mandrillaris at a concentration of 100μM. These combinations included (i) prochlorperazine plus loperamide; (ii) prochlorperazine plus apomorphine; and (iii) procyclidine plus loperamide. In viability assays, none of the drug-treated amoebae emerged as viable trophozoites, suggesting irreversible amoebicidal effects. Four combinations of drugs tested showed varied potency against A. castellanii and B. mandrillaris at 100μM. The combination of haloperidol and loperamide was highly effective against A. castellanii at 100μM, but potent effects against B. mandrillaris were observed only at 250μM. Digoxin and amlodipine were effective against A. castellanii and B. mandrillaris at 100μM and 250μM, respectively. In contrast, the combination of apomorphine and haloperidol was effective against B. mandrillaris and A. castellanii at 100μM and 250μM, respectively. At 100μM, the combination of procyclidine and amiodarone was effective against neither A. castellanii nor B. mandrillaris. In this case, amoebicidal properties were observed at 750μM for A. castellanii, and 950μM for B. mandrillaris. As these drugs are used clinically against non-communicable diseases, the findings reported here have the potential to be tested in a clinical setting against amoebic encephalitis caused by A. castellanii and B. mandrillaris.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
441872
Calcium, dendritic pieces, purified by distillation, 99.99% trace metals basis
Ca
596566
Calcium, dendritic pieces, purified by distillation, 99.9% trace metals basis
Ca
465895
Calcium, dendritic pieces, purified by distillation, 99.5% trace metals basis
Ca
215147
Calcium, granular, 99%
Ca
327387
Calcium, pieces, <1 cm, 99%
Ca
215414
Calcium, turnings, 99% trace metals basis
Ca
04599
Digoxin, certified reference material, TraceCERT®
C41H64O14
D6003
Digoxin, analytical standard
C41H64O14
1200000
Digoxin, United States Pharmacopeia (USP) Reference Standard
C41H64O14
D6770
Digoxin, analytical standard
C41H64O14
D1900000
Digoxin, European Pharmacopoeia (EP) Reference Standard
C41H64O14
Y0000554
Digoxin for peak identification, European Pharmacopoeia (EP) Reference Standard
C41H64O14
H1512
Haloperidol, powder
C21H23ClFNO2
1303002
Haloperidol, United States Pharmacopeia (USP) Reference Standard
C21H23ClFNO2
H100
Haloperidol
C21H23ClFNO2
H0100000
Haloperidol, European Pharmacopoeia (EP) Reference Standard
C21H23ClFNO2
Y0001518
Haloperidol for peak identification, European Pharmacopoeia (EP) Reference Standard
C21H23ClFNO2
Y0001517
Haloperidol for system suitability, European Pharmacopoeia (EP) Reference Standard
C21H23ClFNO2
59964
Histamine, analytical standard
C5H9N3
53290
Histamine, base, ≥97.0% (NT)
C5H9N3