EMAIL THIS PAGE TO A FRIEND
Chemico-biological interactions
Refined biokinetic model for humans exposed to cobalt dietary supplements and other sources of systemic cobalt exposure.
PMID 24726710
Abstract
An updated biokinetic model for human exposures to cobalt (Co) was developed based on a comprehensive set of human pharmacokinetics data collected from five male and five female volunteers who ingested ∼1 mg Co/day of a Co supplement for 3 months. Three key experimental observations from the human dosing studies were incorporated into the model: (1) an increase in the measured fraction of large molecular serum protein bound Co from 95% during dosing to 99% after dosing; (2) a linear decrease in Co red blood cell concentration after dosing; and (3) Co renal clearance consistent with estimated glomerular filtration rates and free Co²⁺ concentration. The model was refined by adding compartments accounting for (1) albumin bound Co in intravascular fluid (serum); (2) albumin bound Co in extravascular fluid with physiologic exchange rates of albumin bound Co between extravascular and intravascular fluid; and (3) a novel sequential cascade of compartments representing red blood cell ages between 1 and 120 days. Reasonable agreement between the modeled and measured urine, serum, and whole blood concentrations were observed (r>0.84, slope=0.79-1.0) with gastrointestinal absorption rates between 9% and 66%. In addition, model predictions agreed well with data from several external studies representing healthy human volunteers, dialysis patients, anephric patients, a Co-poisoning incident and whole body retention studies. Our revised model considerably improves the state of knowledge on human Co kinetics, and should be helpful for evaluating elevated blood Co concentrations in currently exposed populations, such as metal-on-metal (MoM) hip implant patients.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GF94594450
Cobalt, foil, 25mm disks, thickness 0.25mm, as rolled, 99.9%
Co
GF41541476
Cobalt, foil, 25mm disks, thickness 0.25mm, as rolled, 99.99+%
Co
GF45372315
Cobalt, foil, 4mm disks, thickness 0.003mm, 99.9%
Co
GF08560289
Cobalt, foil, 4mm disks, thickness 0.004mm, 99.9%
Co
GF89559343
Cobalt, foil, 4mm disks, thickness 0.005mm, 99.9%
Co
GF48392051
Cobalt, foil, 4mm disks, thickness 0.006mm, 99.9%
Co
GF55954873
Cobalt, foil, 4mm disks, thickness 0.007mm, 99.9%
Co
GF92926727
Cobalt, foil, 4mm disks, thickness 0.008mm, 99.9%
Co
GF70445246
Cobalt, foil, 4mm disks, thickness 0.009mm, 99.9%
Co
GF40089225
Cobalt, foil, 4mm disks, thickness 0.0125mm, 99.9%
Co
GF48813723
Cobalt, foil, 100x100mm, thickness 0.125mm, annealed, 99.9%
Co
GF10782121
Cobalt, foil, 100x100mm, thickness 0.125mm, as rolled, 99.99+%
Co
GF40808516
Cobalt, foil, 100x100mm, thickness 0.25mm, as rolled, 99.9%
Co
GF18093461
Cobalt, foil, 100x100mm, thickness 0.25mm, as rolled, 99.99+%
Co
GF30160291
Cobalt, foil, 100x100mm, thickness 0.5mm, as rolled, 99.99+%
Co
GF81966224
Cobalt, foil, 100x100mm, thickness 0.7mm, annealed, 99.9%
Co
GF56494187
Cobalt, foil, 100x100mm, thickness 1.0mm, as rolled, 99.9%
Co
GF76259593
Cobalt, foil, 100x100mm, thickness 2.0mm, as rolled, 99.9%
Co
GF91564904
Cobalt, foil, 250x250mm, thickness 0.50mm, annealed, 99.9%
Co
GF88288699
Cobalt, foil, 25x25mm, thickness 0.125mm, as rolled, 99.99+%
Co
GF30959411
Cobalt, foil, 25x25mm, thickness 0.25mm, as rolled, 99.9%
Co
GF67617153
Cobalt, foil, 25x25mm, thickness 0.25mm, as rolled, 99.99+%
Co
GF85563535
Cobalt, foil, 25x25mm, thickness 0.5mm, as rolled, 99.99+%
Co
GF09035647
Cobalt, foil, 25x25mm, thickness 0.7mm, annealed, 99.9%
Co
GF41834011
Cobalt, foil, 25x25mm, thickness 1.0mm, as rolled, 99.9%
Co
GF34805268
Cobalt, foil, 25x25mm, thickness 1.0mm, as rolled, 99.99+%
Co
GF40133472
Cobalt, foil, light tested, 100x100mm, thickness 0.05mm, annealed, 99.9%
Co
GF03773640
Cobalt, foil, light tested, 100x100mm, thickness 0.05mm, as rolled, 99.9%
Co
GF06754406
Cobalt, foil, light tested, 100x100mm, thickness 0.05mm, as rolled, 99.99+%
Co
GF42159315
Cobalt, foil, light tested, 25x25mm, thickness 0.009mm, 99.9%
Co