EMAIL THIS PAGE TO A FRIEND
Free radical biology & medicine
Oxidative stress induces claudin-2 nitration in experimental type 1 diabetic nephropathy.
PMID 24726862
Abstract
Renal complications in diabetes are severe and may lead to renal insufficiency. Early alterations in tight junction (TJ) proteins in diabetic nephropathy (DN) have not been explored and the role of oxidative stress in their disassembly has been poorly characterized. We investigated the expression and distribution of TJ proteins: claudin-5 in glomeruli (GL), occludin and claudin-2 in proximal tubules (PTs), and ZO-1 and claudin-1, -4, and -8 in distal tubules (DTs) of rats 21 days after streptozotocin injection. Redox status along the nephron segments was evaluated. Diabetes increased kidney injury molecule-1 expression. Expression of sodium glucose cotransporters (SGLT1 and SGLT2) and facilitative glucose transporter (GLUT2) was induced. Increased oxidative stress was present in GL and PTs and to a lesser extent in DTs (measured by superoxide production and PKCβ2 expression), owing to NADPH oxidase activation and uncoupling of the endothelial nitric oxide synthase-dependent pathway. Claudin-5, occludin, and claudin-2 expression was decreased, whereas claudin-4 and -8 expression increased. ZO-1 was redistributed from membrane to cytosol. Increased nitration of tyrosine residues in claudin-2 was found, which might contribute to decrement of this protein in proximal tubule. In contrast, occludin was not nitrated. We suggest that loss of claudin-2 is associated with increased natriuresis and that loss of glomerular claudin-5 might explain early presence of proteinuria. These findings suggest that oxidative stress is related to alterations in TJ proteins in the kidney that are relevant to the pathogenesis and progression of DN and for altered sodium regulation in diabetes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D6665
2′,7′-Dichlorofluorescein, ~90% (TLC), crystalline
C20H10Cl2O5
35848
2′,7′-Dichlorofluorescein, suitable for fluorescence, BioReagent, ≥90% (T)
C20H10Cl2O5
410217
2′,7′-Dichlorofluorescein, ACS reagent
C20H10Cl2O5
B1378
2,6-Di-tert-butyl-4-methylphenol, ≥99.0% (GC), powder
C15H24O
34750
2,6-Di-tert-butyl-4-methylphenol, purum, ≥99.0% (GC)
C15H24O
03762
2,6-Di-tert-butyl-4-methylphenol, tested according to Ph.Eur.
C15H24O
AV42845
Anti-CLDN2 antibody produced in rabbit, IgG fraction of antiserum
1082708
Butylated hydroxytoluene, United States Pharmacopeia (USP) Reference Standard
C15H24O
W218405
Butylated hydroxytoluene, ≥99%, FCC, FG
C15H24O
PHR1117
Butylated Hydroxytoluene, Pharmaceutical Secondary Standard; Certified Reference Material
C15H24O
C4528
Chlorobimane
C10H11ClN2O2
W381918
L-Arginine, 99%, FCC, FG
C6H14N4O2
A5006
L-Arginine, reagent grade, ≥98%
C6H14N4O2
A8094
L-Arginine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
C6H14N4O2
A4474
L-Arginine, PharmaGrade, Ajinomoto, EP, USP, manufactured under appropriate GMP controls for Pharma or Biopharmaceutical production, suitable for cell culture
C6H14N4O2
PHR1106
L-Arginine, Pharmaceutical Secondary Standard; Certified Reference Material
C6H14N4O2
11009
L-Arginine, BioUltra, ≥99.5% (NT)
C6H14N4O2
69899
Monochlorobimane, suitable for fluorescence, ≥70.0% (HPCE)
C10H11ClN2O2
X0626
Xanthine, ≥99.5% (HPLC), purified by recrystallization
C5H4N4O2
X4002
Xanthine, BioUltra, ≥99%
C5H4N4O2
X7375
Xanthine, ≥99%
C5H4N4O2