EMAIL THIS PAGE TO A FRIEND
Ecotoxicology and environmental safety
Developmental toxicity in rare minnow (Gobiocypris rarus) embryos exposed to Cu, Zn and Cd.
PMID 24726939
Abstract
Using rare minnow (Gobiocypris rarus) embryos as experimental model, the developmental toxicity of Cu, Zn and Cd were investigated following exposure to 0.001-1.000mg/L for 72h, and the toxicity effects were evaluated by larval malformation rate, heart rate, pericardial area, spontaneous movements, tail length, enzyme activities and biomarker genes. Our results revealed that increased malformation rate provide a gradual dose-response relationship, and the most pronounced morphological alteration was heart and body malformations. Values of 72h EC50 with their 95 percent confidence intervals on G. rarus embryos were 0.103 (0.072-0.149)mg/L for Cu, 0.531 (0.330-1.060)mg/L for Zn, 0.219 (0.147-0.351)mg/L for Cd. Enzyme activities can be regard as a type of low-dose stimulation and high-dose inhibition. Stress and metabolism-related genes (hsp70, cyp1a and mt) were significantly up-regulated, development-related genes (wnt8a, vezf1 and mstn) were significantly down-regulated after the treatment by Cu, Zn and Cd. Overall, the present study points out Cu, Zn and Cd are highly toxic to G. rarus embryos. The information presented in this study will be helpful in fully understanding the toxicity induced by Cu, Zn and Cd in fish embryos.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GF61936814
Cadmium, foil, 100x100mm, thickness 0.25mm, as rolled, 99.99+%
Cd
GF17721637
Cadmium, foil, 100x100mm, thickness 0.5mm, as rolled, 99.95%
Cd
GF64286602
Cadmium, foil, 100x100mm, thickness 1.0mm, as rolled, 99.99%
Cd
GF30096345
Cadmium, foil, 100x100mm, thickness 2.0mm, as rolled, 99.99%
Cd
GF10741889
Cadmium, foil, 100x100mm, thickness 3.0mm, as rolled, 99.9%
Cd
GF37649100
Cadmium, foil, 100x200mm, thickness 0.125mm, as rolled, 99.95%
Cd
GF38583287
Cadmium, foil, 100x300mm, thickness 0.125mm, as rolled, 99.95%
Cd
GF50990597
Cadmium, foil, 150x150mm, thickness 0.25mm, as rolled, 99.95%
Cd
GF72757503
Cadmium, foil, 150x150mm, thickness 0.5mm, as rolled, 99.99%
Cd
GF66037754
Cadmium, foil, 150x150mm, thickness 1.0mm, as rolled, 99.95%
Cd
GF76768914
Cadmium, foil, 150x150mm, thickness 3.0mm, as rolled, 99.9%
Cd
GF33512688
Cadmium, foil, 200x200mm, thickness 0.5mm, as rolled, 99.99%
Cd
GF94010453
Cadmium, foil, 200x200mm, thickness 1.0mm, as rolled, 99.99%
Cd
GF90633652
Cadmium, foil, 25x25mm, thickness 0.125mm, as rolled, 99.99+%
Cd
GF06836238
Cadmium, foil, 25x25mm, thickness 0.25mm, as rolled, 99.9%
Cd
GF71408763
Cadmium, foil, 25x25mm, thickness 0.25mm, as rolled, 99.99+%
Cd
GF59435062
Cadmium, foil, 25x25mm, thickness 0.5mm, as rolled, 99.999%
Cd
GF78263411
Cadmium, foil, 25x25mm, thickness 1.0mm, as rolled, 99.999%
Cd
GF84554613
Cadmium, foil, 300x1000mm, thickness 2.0mm, as rolled, 99.99%
Cd
GF19008817
Cadmium, foil, 300x300mm, thickness 0.25mm, as rolled, 99.95%
Cd
GF20958947
Cadmium, foil, 300x300mm, thickness 0.5mm, as rolled, 99.99%
Cd
GF15879954
Cadmium, foil, 300x300mm, thickness 2.0mm, as rolled, 99.99%
Cd
GF72851182
Cadmium, foil, 30x100mm, thickness 0.5mm, as rolled, 99.999%
Cd
GF61336997
Cadmium, wire reel, 5m, diameter 0.5mm, hard, 99.99+%
Cd
GF88672786
Cadmium, wire reel, 5m, diameter 0.125mm, as drawn, 99.9%
Cd
GF97910950
Cadmium, wire reel, 0.1m, diameter 0.125mm, as drawn, 99.9%
Cd
GF42263849
Cadmium, wire reel, 0.1m, diameter 1.0mm, hard, 99.999%
Cd
GF12224078
Cadmium, wire reel, 0.2m, diameter 0.25mm, hard, 99.99+%
Cd
GF23019960
Cadmium, wire reel, 0.2m, diameter 0.5mm, hard, 99.95+%
Cd
GF37213893
Cadmium, wire reel, 0.2m, diameter 0.5mm, hard, 99.99+%
Cd