EMAIL THIS PAGE TO A FRIEND
Journal of Pharmaceutical and Biomedical Analysis
Investigation of a new core-shell particle column for ion-pair reversed-phase liquid chromatography analysis of oligonucleotides.
PMID 24727281