EMAIL THIS PAGE TO A FRIEND
Clinical pharmacology and therapeutics
A current approach to statin intolerance.
PMID 24727470
Abstract
Statins are the first-line pharmacotherapy for cholesterol reduction. Use of these drugs in large randomized clinical trials has consistently shown significant reductions in major vascular events, including death, myocardial infarction, stroke, and coronary revascularization. The updated guidelines for the treatment of high blood cholesterol from the American College of Cardiology/American Heart Association (ACC/AHA), will lead to an increase in the number of patients taking statins. Hence, the number of cases of statin intolerance may subsequently increase, emphasizing the need to understand and treat this important problem.