EMAIL THIS PAGE TO A FRIEND
Biomedical chromatography : BMC
Therapeutic monitoring of amphotericin B in Saudi ICU patients using UPLC MS/MS assay.
PMID 24733605
Abstract
Amphotericin B (AmB) is the first-line agent for the treatment of life-threatening invasive fungal infections. The aim of this study was to monitor AmB in critically ill Saudi patients in ICU after i.v. administration of 0.68 ± 0.1 mg/kg/day Fungizone®. A selective, sensitive and precise UPLC MS/MS method was developed to measure AmB concentrations in these patients. Seven ICU patients with creatinine clearance (ClCr) >40 mL/min were included. AmB levels were analyzed using a Waters Aquity UPLC MS/MS system, a BEH Shield RP18 column and detection via electrospray ionization source with positive ionization mode. The precision and accuracy of the developed UPLC method in the concentration range of 200-4000 ng/mL show no significant difference among inter- and-intra-day analysis (p > 0.05). Linearity was observed over the investigated range with correlation coefficient, r > 0.995 (n = 6/day). The pharmacokinetics of AmB in these patients, at steady state, showed a high terminal half-life of 124.6 ± 73.4 h, with a highest concentration of 513.9 ± 281.1 ng/mL, a lowest concentration 316.4 ± 129.0 ng/mL and a mean clearance 91.1 ± 39.2 mL/h/kg. The pharmacokinetics of AmB in critically ill Saudi patients in ICU was studied using a fully validated assay. A weak correlation (r = -0.22) of AmB Cl with ClCr was obtained, which suggests the need for further investigation in a larger population.