EMAIL THIS PAGE TO A FRIEND
Journal of nanoscience and nanotechnology
Novel chitosan coated magnetic nanocarriers for the targeted Diclofenac delivery.
PMID 24734550
Abstract
New magnetic devices consisting of magnetite functionalized with oleic acid and chitosan have been synthesized and employed to the loading of Diclofenac as potential tool for treatment of targeted inflammatory diseases. Magnetic loaded and un-loaded nanoparticles have been thoroughly characterized by infrared spectroscopy, transmission electron microscopy, determination of hydrodynamic diameter by Dynamic light scattering and zeta potential measurements at different pH conditions. A study of the release of Diclofenac has been performed in vitro and available mathematical models have been used to determine the release kinetic. Both properties and release data reveal that this nanomagnetic platform would be suitable for in vivo assays.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D1039990 Diclofenac potassium, European Pharmacopoeia (EP) Reference Standard
C14H10Cl2KNO2