EMAIL THIS PAGE TO A FRIEND
Scandinavian Journal of Clinical and Laboratory Investigation
Ultrastructural characterization of mesenchymal stromal cells labeled with ultrasmall superparamagnetic iron-oxide nanoparticles for clinical tracking studies.
PMID 24734781