EMAIL THIS PAGE TO A FRIEND
Biochemical pharmacology
Brazilin selectively disrupts proximal IL-1 receptor signaling complex formation by targeting an IKK-upstream signaling components.
PMID 24735611
Abstract
The ligation of interleukin-1 receptor (IL-1R) or tumor necrosis factor receptor 1 (TNFR1) induces the recruitment of adaptor proteins and their concomitant ubiquitination to the proximal receptor signaling complex, respectively. Such are upstream signaling events of IKK that play essential roles in NF-κB activation. Thus, the discovery of a substance that would modulate the recruitment of key proximal signaling elements at the upstream level of IKK has been impending in this field of study. Here, we propose that brazilin, an active compound of Caesalpinia sappan L. (Leguminosae), is a potent NF-κB inhibitor that selectively disrupts the formation of the upstream IL-1R signaling complex. Analysis of upstream signaling events revealed that brazilin markedly abolished the IL-1β-induced polyubiquitination of IRAK1 and its interaction with IKK-γ counterpart. Notably, pretreatment of brazilin drastically interfered the recruitment of the receptor-proximal signaling components including IRAK1/4 and TRAF6 onto MyD88 in IL-1R-triggerd NF-κB activation. Interestingly, brazilin did not affect the TNF-induced RIP1 ubiquitination and the recruitment of RIP1 and TRAF2 to TNFR1, suggesting that brazilin is effective in selectively suppressing the proximal signaling complex formation of IL-1R, but not that of TNFR1. Moreover, our findings suggest that such a disruption of IL-1R-proximal complex formation by brazilin is not mediated by affecting the heterodimerization of IL-1R and IL-1RAcP. Taken together, the results suggest that the anti-IKK activity of brazilin is induced by targeting IKK upstream signaling components and subsequently disrupting proximal IL-1 receptor signaling complex formation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1406018
Anti-IL1R1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
I8028 Anti-IRAK antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
I0785
Anti-IRAK1 (506-520) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB1406040
Anti-IRAK1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB4501559
Anti-IRAK1 antibody produced in rabbit, affinity isolated antibody
SAB4300304
Anti-REL (Ab-503) antibody produced in rabbit, affinity isolated antibody
SAB2501618
Anti-REL antibody produced in goat, affinity isolated antibody, buffered aqueous solution
PRS3877
Anti-TANK (ab1) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2900286
Anti-TANK (AB1) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
PRS3879
Anti-TANK (ab2) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2900392
Anti-TANK (AB2) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2103894
Anti-TRADD antibody produced in rabbit, affinity isolated antibody
SAB1411558
Anti-TRADD antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB2501057
Anti-TRAF2 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA009972
Anti-TRAF2 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA010634
Anti-TRAF2 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB3500375
Anti-TRAF2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1406553
Anti-TRAF6 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA020599
Anti-TRAF6 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2
HPA019805
Anti-TRAF6 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1
SAB3500377
Anti-TRAF6 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB3500129
Monoclonal Anti-IRAK antibody produced in mouse, clone 8F1A7, purified immunoglobulin, buffered aqueous solution
WH0003654M1
Monoclonal Anti-IRAK1 antibody produced in mouse, clone 3F7, purified immunoglobulin, buffered aqueous solution
WH0008717M3
Monoclonal Anti-TRADD antibody produced in mouse, clone 3G3, purified immunoglobulin, buffered aqueous solution
SAB1404491
Monoclonal Anti-TRAF2 antibody produced in mouse, clone 4C11, ascites fluid
SAB1402384
Monoclonal Anti-TRAF2 antibody produced in mouse, clone 4C11, purified immunoglobulin, buffered aqueous solution
WH0007189M2
Monoclonal Anti-TRAF6 antibody produced in mouse, clone 1B2, purified immunoglobulin, buffered aqueous solution
T7539
Tumor Necrosis Factor-α from mouse, TNF-α, recombinant, expressed in E. coli, powder, suitable for cell culture
T5944
Tumor Necrosis Factor-α from rat, TNF-α, recombinant, expressed in E. coli, powder, suitable for cell culture