EMAIL THIS PAGE TO A FRIEND
Cancer research
Activation of SOX2 expression by BRD4-NUT oncogenic fusion drives neoplastic transformation in NUT midline carcinoma.
PMID 24736545
Abstract
BRD4 is implicated in the pathogenesis of a number of different cancers. It is also the target of translocation t(15;19) that accounts for the highly aggressive NUT midline carcinoma (NMC). We discovered that t(15;19) NMC cells display the ability to grow into stem cell-like spheres and express an exceptionally high level of the stem cell marker, SOX2. The BRD4-NUT fusion oncogene resulting from t(15;19) translocation is required for the abnormal activation of SOX2, which drives the stem cell-like proliferation and cellular transformation in NMC cells. SOX2 knockdown phenocopies the effects of BRD4-NUT inhibition, whereas ectopic SOX2 expression rescues the phenotype. The BRD4-NUT-induced abnormal SOX2 activation was observed in multiple NMC cell lines as well as in NMC primary tumors. We further demonstrate that BRD4-NUT oncoprotein recruits p300 to stimulate transcription activation and that inhibition of p300 represses SOX2 transcription in NMC cells. These studies identify this stem cell marker as a novel BRD4-NUT target that supports the highly aggressive transforming activity of t(15;19) carcinomas. Our study provides new mechanistic insights for understanding how alteration of BRD4 function by BRD4-NUT oncogene leads to the highly malignant NMC carcinoma. Because abnormal stem cell self-renewal is frequently observed during tumor formation and metastasis, the aberrant stem cell-like proliferation associated with BRD4 dysregulation observed in NMC carcinoma may have implications for studying the oncogenic mechanism of other BRD4-associated tumors.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
56454
1-Thioglycerol, PharmaGrade, USP, Manufactured under appropriate GMP controls for pharma or biopharmaceutical production.
C3H8O2S
M1753
1-Thioglycerol, ≥97%
C3H8O2S
M6145
1-Thioglycerol, liquid, BioReagent, suitable for cell culture, ≥97% (titration)
C3H8O2S
M2172
1-Thioglycerol, BioXtra, ≥97%
C3H8O2S
88640
1-Thioglycerol, ≥99.0% (GC)
C3H8O2S
SAB2701800
Anti-sox2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
S9072 Anti-SOX2 antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
AV38232
Anti-SOX2 antibody produced in rabbit, IgG fraction of antiserum
SAB3500363
Anti-SOX2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2104660
Anti-SOX2 antibody produced in rabbit, affinity isolated antibody
SAB4300669
Anti-SOX2 antibody produced in rabbit, affinity isolated antibody
SAB2701973
Anti-SOX2 antibody produced in rabbit
SAB2701974
Anti-SOX2 antibody produced in rabbit
SAB2103958
Anti-SOX2, (N-terminal) antibody produced in rabbit, affinity isolated antibody
EHU184131 MISSION® esiRNA, esiRNA targeting human Sox2
RDD004
PIPES, anhydrous, free-flowing, Redi-Dri, ≥99%
C8H18N2O6S2
P6757
PIPES, ≥99% (titration)
C8H18N2O6S2
RES0703P-A7
PIPES, PharmaGrade, Manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production, suitable for cell culture
C8H18N2O6S2
P1851
PIPES, BioPerformance Certified, suitable for cell culture, ≥99%
C8H18N2O6S2
P8203
PIPES, BioXtra, ≥99% (titration)
C8H18N2O6S2
80635
PIPES, BioXtra, for molecular biology, ≥99.5% (T)
C8H18N2O6S2